Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-049

Supelco

Psilocin-D10

vial of 5 mg, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6N2OD10
Molecular Weight:
214.33
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 5 mg

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

A stable-labeled Certified Reference Material suitable for quantification of Psilocin levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Psilocin is a potent hallucinogen present in most psychedelic mushrooms. Known by street names, Shrooms, Boomers, or Musk, psilocin has effects similar to LSD and DMT.

Legal Information

Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service