Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L-004

Supelco

LAMPA solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Lysergic acid N,N-methylpropylamide, Lysergic acid N,N-methylpropylamide solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

CCCN(C)C(=O)[C@H]1CN(C)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34

InChI

1S/C20H25N3O/c1-4-8-22(2)20(24)14-9-16-15-6-5-7-17-19(15)13(11-21-17)10-18(16)23(3)12-14/h5-7,9,11,14,18,21H,4,8,10,12H2,1-3H3/t14-,18-/m1/s1

InChI key

CZRJGQXHVRNZRZ-RDTXWAMCSA-N

General description

LAMPA, or lysergic acid methyl propyl amide, is an isomer of the illicit psychedelic drug LSD, also known as lysergic acid diethylamide. This analytical reference standard is suitable for LSD/LAMPA differentiation cOnfi®rmation in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Onfi is a registered trademark of Lundbeck LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service