Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F-028

Supelco

(±)-4-Fluoroephedrine-D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11D3FNO · HCl
Molecular Weight:
222.70
EC Number:

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

concentration

100 μg/mL in methanol (as free base)

storage temp.

−20°C

SMILES string

Cl.[2H]C([2H])([2H])NC(C)C(O)c1ccc(F)cc1

General description

This bath salt cathinone analog is a primary urinary metabolite of 4-Fluoromethcathinone. 4-Fluoromethcathinone, also known as flephedrone or 4-FMC, is a synthetic cathinone and stimulant widely found in numerous bath salt blends and party pills. This certified Snap-N-Spike® solution is suitable as an internal standard for quantitation of 4-Fluoroephedrine by LC/MS or GC/MS for applications in urine drug testing, clinical toxicology, or forensic analysis.

Legal Information

Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service