Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-114

Supelco

N-Desmethyltrimipramine-D3 hydrochloride solution

100 μg/mL in methanol (as free base), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21D3N2 · HCl
Molecular Weight:
319.89
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

CC(CNC([2H])([2H])[2H])CN1C2=C(C=CC=C2)CCC3=C1C=CC=C3.[H]Cl

InChI

1S/C19H24N2.ClH/c1-15(13-20-2)14-21-18-9-5-3-7-16(18)11-12-17-8-4-6-10-19(17)21;/h3-10,15,20H,11-14H2,1-2H3;1H/i2D3;

InChI key

LRSZUGQWBGGCTG-MUTAZJQDSA-N

General description

N-Desmethyltrimipramine is a primary blood metabolite of trimipramine. Trimipramine is a tricyclic antidepressant prescribed for treatment of depression, insomnia, and chronic pain or as an adjunctive therapy for alcohol or opioid withdrawal. This stable-labeled internal standard is suitable for quantification of N-Desmethyltrimipramine levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service