Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B-057

Supelco

(±)-Bromo-DragonFLY-D5 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7D5BrNO2 · HCl
Molecular Weight:
335.64
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

Bromo-DragonFLY is an analog of the 2C amine family that also includes 2C-B, 2C-I, and 2C-E. This designer drug is an extremely potent hallucinogen with duration of action much longer than LSD. Toxicity of Bromo-DragonFLY appears to be fairly high for humans when taken in larger doses. This stable-labeled internal standard is suitable for quantitation of Bromo-DragonFLY levels in urine, serum, or plasma by GC/MS or LC/MS methods for use in clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service