Key Documents
229452
CELLTREAT
CELLTREAT Centrifuge tubes
TUBE & CAP, 15mL Centrifuge Tube & Cap - Bags, Non-sterile (Tubes and Caps Packed Separately)
Synonym(s):
Centrifuge tube caps
About This Item
PDF Icon
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
CELLTREAT
General description
  • Centrifuge Tubes and Caps packed separately
  • Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
  • Large white marking area provides excellent contrast for identification
  • Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
  • Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
Choose from one of the most recent versions:
Certificates of Analysis (COA)