Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1157-0020

CELLTREAT

VistaLab Pipettes

20µL Pipette, Ovation, (F1) Fixed Volume, One Stroke, Yellow

Synonym(s):

Pipettors

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$218.40

$218.40


In StockDetails



Select a Size

Change View
1 EA
$218.40

About This Item

PDF Icon
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
PDF Icon
Have a question?

$218.40


In StockDetails


manufacturer/tradename

CELLTREAT

General description

Ovation® pipettes have a unique and patented ergonomic design. They are the only pipettes that fully address the issues of posture, repetition, and force to significantly reduce strain and pain that occur from pipetting, while always providing superior accuracy and precision.

The angled shape allows users to pipette with low arm and elbow height, decreasing tension and associated pain in the neck, shoulder, back, and elbow. When used properly, the Ovation pipette will keep the user's hand and wrist in the neutral position recommended by ergonomic experts.

Contoured body shape and adjustable hook assures a comfortable, easy grip, and custom fit hand hold.

Spring-loaded tip acquisition means a gentle push is all that is required to attach tips, and an audible "click" confirms secure attachment.

Tips "flick" off easily with a low force push of the eject button.

Special filters in the nozzle prevent liquid from being inadvertently aspirated into the pipettor, protecting it from contamination.

Ovation pipettes are the only pipettes that stand on their own, which means tips never touch the bench, even if the pipette is put down in the middle of a procedure.

Mechanical models feature two-stroke operation, available with "click stop" or "quick set digital" volume adjustment (0.2µL - 1000µL).

Electronic models feature custom volume presets, multi-dispensing, mixing, and serial diluting (0.5µL - 1250µL).

Electronic Macro 5mL and 10mL, and electronic 8 and 12 multichannel models available.

Single channel models include a built-in micro tube cap opener.

Color-coded for easy and quick volume identification.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library