Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860642C

Avanti

Dimethyl Sphingosine (d17:1)

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

N,N-dimethyl-D-erythro-sphingosine (C17 base)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H39NO2
CAS Number:
Molecular Weight:
313.52
UNSPSC Code:
12352211
NACRES:
NA.25

assay

99% (TLC)

form

chloroform solution

packaging

1 ea of 5 mg (860642C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N(C)C)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCC

General description

Dimethyl Sphingosine is a derivative of sphingosine. It is found in many tissues.[1]

Biochem/physiol Actions

Dimethyl Sphingosine plays a major role in the inhibition of protein kinase C (PKC) and sphingosine kinase. It is also involved in the inhibition of src kinase and augmentation of epidermal growth factor receptor (EGFR).[2]

Packaging

5 mL Clear Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N, N-Dimethyl-d-erythro-sphingosine inhibits store-operated Ca2+ entry in U937 monocytes
Jo J Y, et al.
Journal of Pharmacological Sciences, 107(3), 303-307 (2008)
Characterization of N, N,-dimethyl-d-erythro-sphingosine-induced apoptosis and signaling in U937 cells: Independence of sphingosine kinase inhibition
Kim H L, et al.
Prostaglandins & other lipid mediators, 86(1-4), 18-25 (2008)
Tania C B Santos et al.
Langmuir : the ACS journal of surfaces and colloids, 36(21), 6007-6016 (2020-05-06)
Compared to the canonical sphingoid backbone of sphingolipids (SLs), atypical long-chain bases (LCBs) lack C1-OH (1-deoxy-LCBs) or C1-CH2OH (1-deoxymethyl-LCBs). In addition, when unsaturated, they present a cis-double bond instead of the canonical  Δ4-5 trans-double bond. These atypical LCBs are directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service