Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATC429238240

Atlantic Research Chemicals Ltd

3-Amino-5-bromo-2-chlorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
250.48
MDL number:

Iupac Name

3-amino-5-bromo-2-chlorobenzoic acid

Inchi Code

1S/C7H5BrClNO2/c8-3-1-4(7(11)12)6(9)5(10)2-3/h1-2H,10H2,(H,11,12)

InChI key

LXCCUZBBWURDLS-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=CA001641

MSDS

https://atlantic-chemicals.com/product/?code=CA001641


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library