Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATC311765800

Atlantic Research Chemicals Ltd

Ethyl Difluoro(6-methoxypyridin-2-yl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
231.2
MDL number:
Product ATC311765800 is currently not available for purchase. Contact Technical Service

Iupac Name

ethyl difluoro(6-methoxy-2-pyridinyl)acetate

Inchi Code

1S/C10H11F2NO3/c1-3-16-9(14)10(11,12)7-5-4-6-8(13-7)15-2/h4-6H,3H2,1-2H3

InChI key

FHGUGEYURXDXDX-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=ES00279

MSDS

https://atlantic-chemicals.com/product/?code=ES00279


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library