Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH98B84BBE

AstaTech, Inc.

3-IODO-4-METHYL-4H-1,2,4-TRIAZOLE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
208.99
MDL number:
Product ATEH98B84BBE is currently not available for purchase. Contact Technical Service

Iupac Name

3-iodo-4-methyl-4H-1,2,4-triazole

Inchi Code

1S/C3H4IN3/c1-7-2-5-6-3(7)4/h2H,1H3

InChI key

OAVQTXRIHZMZFV-UHFFFAOYSA-N

Purity

95%

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=147475

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=98649


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library