Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH961B8E27

AstaTech, Inc.

TERT-BUTYL 4-OXO-9-AZADISPIRO[2.1.25.33]DECANE-9-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
251.33
MDL number:
Product ATEH961B8E27 is currently not available for purchase. Contact Technical Service

Iupac Name

tert-butyl 4-oxo-9-azadispiro[2.1.2(5).3(3)]decane-9-carboxylate

Inchi Code

1S/C14H21NO3/c1-12(2,3)18-11(17)15-8-13(4-5-13)10(16)14(9-15)6-7-14/h4-9H2,1-3H3

InChI key

RSOXVBNWRSTZPL-UHFFFAOYSA-N

Purity

95%

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=165868

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W19662


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library