Skip to Content
Merck
All Photos(1)

Key Documents

ATEH03AB6143

AstaTech, Inc.

(4-BROMOPHENYL)(PYRIDIN-2-YL)METHANAMINE HCL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
299.6
MDL number:

Pricing and availability is not currently available.

Iupac Name

(4-bromophenyl)(pyridin-2-yl)methanamine hydrochloride

Inchi Code

1S/C12H11BrN2.ClH/c13-10-6-4-9(5-7-10)12(14)11-3-1-2-8-15-11;/h1-8,12H,14H2;1H

InChI key

NNERKGVXQBQKNE-UHFFFAOYSA-N

Purity

95%

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=116514

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F19202


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library