Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH03AB3FB9

AstaTech, Inc.

TERT-BUTYL 2-AMINO-4,5,7,8-TETRAHYDROTHIAZOLO[5,4-D]AZEPINE-6-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
269.37
MDL number:
Product ATEH03AB3FB9 is currently not available for purchase. Contact Technical Service

Iupac Name

tert-butyl 2-amino-4,5,7,8-tetrahydro-6H-thiazolo[4,5-d]azepine-6-carboxylate

Inchi Code

1S/C12H19N3O2S/c1-12(2,3)17-11(16)15-6-4-8-9(5-7-15)18-10(13)14-8/h4-7H2,1-3H3,(H2,13,14)

InChI key

CIEFBAXZZCPNLH-UHFFFAOYSA-N

Purity

95%

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=114708

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=S11758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library