ATE443546977
AstaTech, Inc.
TERT-BUTYL 3-FLUORO-4-OXOAZEPANE-1-CARBOXYLATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl 3-fluoro-4-oxo-1-azepanecarboxylate
Inchi Code
1S/C11H18FNO3/c1-11(2,3)16-10(15)13-6-4-5-9(14)8(12)7-13/h8H,4-7H2,1-3H3
InChI key
IDKJZSHWZNDJJV-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
95%
Shipping Temperature
Normal
Physical Form
Sticky Oil to Semi-Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=55190
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=72172
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.