Skip to Content
Merck
All Photos(1)

Key Documents

ATE429353365

AstaTech, Inc.

ETHYL 4,5,6,7-TETRAHYDROISOXAZOLO[4,5-C]PYRIDINE-3-CARBOXYLATE HCL

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
232.67
MDL number:

Iupac Name

ethyl 4,5,6,7-tetrahydroisoxazolo[4,5-c]pyridine-3-carboxylate hydrochloride

Inchi Code

1S/C9H12N2O3.ClH/c1-2-13-9(12)8-6-5-10-4-3-7(6)14-11-8;/h10H,2-5H2,1H3;1H

InChI key

SLAKBPPVIPODKW-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=49821

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=89039


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library