Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE428005189

AstaTech, Inc.

2,3-DICHLOROQUINOXALINE-6-CARBOXYLIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
243.05
MDL number:
Product ATE428005189 is currently not available for purchase. Contact Technical Service

Iupac Name

2,3-dichloro-6-quinoxalinecarboxylic acid

Inchi Code

1S/C9H4Cl2N2O2/c10-7-8(11)13-6-3-4(9(14)15)1-2-5(6)12-7/h1-3H,(H,14,15)

InChI key

ZGQQEHVULTVXQD-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=39551

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=27924


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library