Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE389310553

AstaTech, Inc.

TERT-BUTYL 2-(4-BROMO-2-CHLOROPHENYL)MORPHOLINE-4-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
376.68
MDL number:
Product ATE389310553 is currently not available for purchase. Contact Technical Service

Iupac Name

tert-butyl 2-(4-bromo-2-chlorophenyl)morpholine-4-carboxylate

Inchi Code

1S/C15H19BrClNO3/c1-15(2,3)21-14(19)18-6-7-20-13(9-18)11-5-4-10(16)8-12(11)17/h4-5,8,13H,6-7,9H2,1-3H3

InChI key

NNHIZVJTGUMGOW-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=17436

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=81353


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library