APOH11C5AE8F
Apollo Scientific Ltd
3-(Piperidin-4-yl)pyridine dihydrochloride
About This Item
Iupac Name
3-(piperidin-4-yl)pyridine dihydrochloride
Inchi Code
1S/C10H14N2.2ClH/c1-2-10(8-12-5-1)9-3-6-11-7-4-9;;/h1-2,5,8-9,11H,3-4,6-7H2;2*1H
Inchi Key
PUOFTXQLJPYTRT-UHFFFAOYSA-N
storage temp.
Country of Origin
Shipping Temperature
Physical Form
Product Link
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.