Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO456796945

Apollo Scientific Ltd

3-Fluoro-5-(trifluoromethyl)phenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
222.14
MDL number:
Product APO456796945 is currently not available for purchase. Contact Technical Service

bp

110-112

Iupac Name

[3-fluoro-5-(trifluoromethyl)phenyl]acetic acid

Inchi Code

1S/C9H6F4O2/c10-7-2-5(3-8(14)15)1-6(4-7)9(11,12)13/h1-2,4H,3H2,(H,14,15)

InChI key

LQIBHDUPSIQRPV-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/3-fluoro-5-trifluoromethylphenylacetic-acid

Synonym

3-(Carboxymethyl)-5-fluorobenzotrifluoride, 2-[3-Fluoro-5-(trifluoromethyl)phenyl]ethanoic acid

Boiling point

110-112

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-fluoro-5-trifluoromethylphenylacetic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library