Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455838561

Apollo Scientific Ltd

2-Amino-4-fluoropyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
112.11
MDL number:
Product APO455838561 is currently not available for purchase. Contact Technical Service

Iupac Name

4-fluoro-2-pyridinamine

Inchi Code

1S/C5H5FN2/c6-4-1-2-8-5(7)3-4/h1-3H,(H2,7,8)

InChI key

WCCFLOQQACDOAX-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/2-amino-4-fluoropyridine

Synonym

4-Fluoropyridin-2-amine

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-amino-4-fluoropyridine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library