APO428499235
Apollo Scientific Ltd
4-Butoxy-2,6-difluorobenzonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-butoxy-2,6-difluorobenzonitrile
Inchi Code
1S/C11H11F2NO/c1-2-3-4-15-8-5-10(12)9(7-14)11(13)6-8/h5-6H,2-4H2,1H3
InChI key
DRMAQHPUVUFLAE-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
97%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/4-butoxy-26-difluorobenzonitrile
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.