Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO428498826

Apollo Scientific Ltd

3,4-Dichloro-5-(trifluoromethyl)benzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
307.92
MDL number:
Product APO428498826 is currently not available for purchase. Contact Technical Service

Iupac Name

5-(bromomethyl)-1,2-dichloro-3-(trifluoromethyl)benzene

Inchi Code

1S/C8H4BrCl2F3/c9-3-4-1-5(8(12,13)14)7(11)6(10)2-4/h1-2H,3H2

InChI key

NTYHLYVDLXQFAO-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/34-dichloro-5-trifluoromethylbenzyl-bromide

pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/34-dichloro-5-trifluoromethylbenzyl-bromide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library