Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AOBH3CD9F246

AOBChem

Methyl 5-acetyl-2-methoxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
208.21
MDL number:
Product AOBH3CD9F246 is currently not available for purchase. Contact Technical Service

Iupac Name

methyl 5-acetyl-2-methoxybenzoate

Inchi Code

1S/C11H12O4/c1-7(12)8-4-5-10(14-2)9(6-8)11(13)15-3/h4-6H,1-3H3

InChI key

KPDRDAXDDZAPHS-UHFFFAOYSA-N

Purity

97

Country of Origin

CN

Physical Form

Solid

Product Link

https://aobchem.com/methyl-5-acetyl-2-methoxybenzoate.html

MSDS

https://aobchem.com/pub/media/files/sds/35812.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library