Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AOBH042EEAB8

AOBChem

2-Hydroxy-6-trifluoromethylphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
205.93
MDL number:
Product AOBH042EEAB8 is currently not available for purchase. Contact Technical Service

Iupac Name

2-hydroxy-6-(trifluoromethyl)phenylboronic acid

Inchi Code

1S/C7H6BF3O3/c9-7(10,11)4-2-1-3-5(12)6(4)8(13)14/h1-3,12-14H

InChI key

FNBWZLJDEFUGAL-UHFFFAOYSA-N

Purity

97

Country of Origin

CN

Physical Form

Solid

Product Link

https://aobchem.com/2-hydroxy-6-trifluoromethylphenylboronic-acid.html

MSDS

https://aobchem.com/pub/media/files/sds/34193.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library