Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH93EDD2DA

Ambeed, Inc.

Ethyl 2,2-Difluoropropionate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
138.11
MDL number:

Iupac Name

ethyl 2,2-difluoropropanoate

Inchi Code

1S/C5H8F2O2/c1-3-9-4(8)5(2,6)7/h3H2,1-2H3

Inchi Key

ISVLDAIKRGXNCZ-UHFFFAOYSA-N

Storage temp.

Sealed in dry,2-8C

Purity

98%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

liquid

Product Link

https://www.ambeed.com/products/28781-85-3.html

Pictograms

Exclamation markFlame

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/28781-85-3.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library