Key Documents
TS100
Alkali Scientific™
Alkali Scientific™ Cell Strainer
100 um, PP, Gamma Sterilized, 50/Pk
Select a Size
1 EA
$74.95
$74.95
Available to ship onApril 01, 2025Details
Select a Size
Change View
1 EA
$74.95
About This Item
PDF Icon
Shipped & Sold By Alkali Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?
$74.95
Available to ship onApril 01, 2025Details
manufacturer/tradename
Alkali Scientific™
General description
Cell Strainers make obtaining a more uniform single cell suspension easy. Three nylon mesh pore sizes are available for isolating cells of different sizes, and cell strainers are faster and eAlkali er than gauze filtering to remove cell clumps and tissues that interfere with sensitive instrumentation. Ideal for stem cell and tissue-derived primary cell preparation. Tab enables easy aseptic handling. Individually packaged and gamma sterilized. Cell strainers from Alkali are an economical choice. Easy to use and simple to dispose of after use, these bAlkali c yet highly functional devices can be eAlkali ly fitted onto standard conical tubes as well as other types of laboratory test tubes with the goal of isolating certain types of cells and of breaking clusters of cells or tissues in single cells in order to produce uniform, single cell suspensions. Thus, they provide a much faster alternative, which at the same time requires less effort on behalf of the researcher, to gauze filtration. Additionally, using strainers will help you to lower the risk of contamination for your sample to a minimum, ensuring that it maintains its integrity. These cell strainers are manufactured from a strong, very resistant nylon mesh which has evenly spaced pores on its surface, providing a uniform filtration platform which will ensure constant, good results. You can choose from three available mesh sizes, which are 40 µm, 70 µm and 100 µm, the different pore sizes help you to isolate different sized cells. Our nylon mesh cell strainers feature an extended molded lip which allows you to eAlkali ly handle the strainer while keeping it by using a forceps or any other laboratory instrument. They have standard dimensions, meaning they can be used to a variety of test tubes commonly found in laboratories. Depending on the size of their pores, these cell filters will provide you and your lab with optimal results in a variety of tasks, by isolating cells of different sizes, breaking up clusters of cells or tissue and keeping debris away. They are ideal for preparing specimens for flow cytometry, because of their ability of removing cell clumps and foreign objects from cell suspensions and research samples before the analyses, which will protect expensive and sensitive instruments such as the flow cytometer or the cell sorter. These cell strainers are also used for primary cell culture, as well as in procedures involving stem cells, cells derived from tissues and cancer cells, such as the operations of preparing primary cell cultures and immunogens. The nylon mesh cell strainers from Alkali are produced in carefully controlled conditions, by highly trained staff using state of the art technology which will test the quality of the products throughout the manufacturing process. The raw material used for the production of these items is high quality, research grade, non-cytotoxic nylon mesh, is made from high quality nylon. This material has high abrAlkali on and alkali resistance, and a high resistance to pressure. The softening point of the material is at around 210° C and the melting point of the nylon mesh is at about 230° C. The fact that the material is non-cytotoxic is a big asset, as it means it will not interfere with the quality of your sample and that it will not kill the cells within it. In order to ensure the success of your research, each strainer we ship is pre-sterilized (SAL 10^-6 sterilization) with the use of low-level gamma radiation (gamma irradiation) after being individually wrapped in medical grade packaging that will ensure it′s ready to use as soon as you pull it out of the box. Cell strainers are packaged in quantities of 50 pieces per pack. For more information on cell strainers view the following videos: Cell Strainer Information Learn more about 70 um cell strainers
Choose from one of the most recent versions:
Certificates of Analysis (COA)