Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DC0077

Alkali Scientific™

Alkali Scientific™ CellPro™ LB (Lennox) + Ampicillin 100ug/mL w/ 2.0% Agar

100mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$44.95

$44.95


Available to ship onFebruary 28, 2025Details



Select a Size

Change View
1 EA
$44.95

About This Item

PDF Icon
Shipped & Sold By Alkali Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?

$44.95


Available to ship onFebruary 28, 2025Details


manufacturer/tradename

Alkali Scientific™

General description

CellPro™ LB (Lennox) + Ampicillin 100ug/mL w/ 2.0% Agar 100mm

Legal Information

CellPro™ is a trademark of Alkali Scientific.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library