Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z417041

Sigma-Aldrich

Aldrich® System 45 open-top caps

diam. 45 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$118.00

$118.00


Available to ship onApril 22, 2025Details



Select a Size

Change View
1 PKG
$118.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$118.00


Available to ship onApril 22, 2025Details


packaging

pkg of 10

diam.

45 mm

Looking for similar products? Visit Product Comparison Guide

General description

Caps are PBT. Temp. range: -45 to 180°C. For use with PTFE-faced silicone liners listed below. Use with syringes for reagent introduction or sampling.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service