Key Documents
Z162124
Water-jet aspirator pump
With pressure gauge
About This Item
UNSPSC Code:
41100000
material
polypropylene
manufacturer/tradename
Azlon
General description
Side-arm tapers from 7 to 12 mm with an integral check valve. Operates effectively (half volume) at a low flow rate of 7 L/min. Max. vacuum is 25 mm Hg (abs) at a flow rate of 17 L/min and a water pressure of 2.8 kg/cm2.
comparable product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)