Key Documents
Z155292
Stainless steel pan Type 304
heavy weight, L × W × H 12.75 in. × 6.9 in. × 2.5 in.
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
stainless steel container
manufacturer/tradename
Polarware S12062
L × W × H
12.75 in. × 6.9 in. × 2.5 in.
capacity
2.25 qt
General description
Wide assortment of standard and heavy duty, seamless trays have coved corners for easy sterilization. Covers have recessed strap handles for ease in handling and storing.
Supplied without cover.
Choose from one of the most recent versions:
Certificates of Analysis (COA)