Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W392405

Sigma-Aldrich

cis-2-Hexen-1-ol

≥96%, stabilized, FG

Synonym(s):

(Z)-2-Hexen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH2OH
CAS Number:
Molecular Weight:
100.16
FEMA Number:
3924
Beilstein/REAXYS Number:
1719708
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.156
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

vapor density

>1 (vs air)

assay

≥96%

contains

α-tocopherol as stabilizer

refractive index

n20/D 1.441 (lit.)

bp

166 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

fresh; green; vegetable; wine-like

SMILES string

[H]\C(CO)=C(/[H])CCC

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h4-5,7H,2-3,6H2,1H3/b5-4-

InChI key

ZCHHRLHTBGRGOT-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-2-Hexen-1-ol is a green leaf volatile (GLV) that is commonly found in herbaceous plants and angiosperm trees.[1] In combination with other GLVs, cis-2-hexen-1-ol may disrupt the response of conifer-infesting ambrosia beetle to the host pheromone and kairomones. This ability makes it a promising candidate for the development of log protectants against ambrosia beetles.[2]

Biochem/physiol Actions

Odor at 1%
Taste at 5-50 ppm

Other Notes

Natural occurrence: Fresh and cooked apple, sour cherry, blueberry, black currant, papaya, peach, french fried potato, tomato, butter, hop oil, cognac, red and white wines, and black tea.
May contain 0.10% E307 synthetic alpha tocopherol

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

138.9 °F - closed cup

flash_point_c

59.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green leaf volatiles disrupt and enhance response to aggregation pheromones by the ambrosia beetle, Gnathotrichus sulcatus (Coleoptera: Scolytidae).
Deglow EK & Borden JH.
Canadian Journal of Forest Research, 28(11), 1697-1705 (1998)
Mosciano, G.
Perfumer & Flavorist, 25, 74-74 (2000)
Christopher J Fettig et al.
Journal of economic entomology, 98(6), 2041-2048 (2006-03-17)
Nonhost angiosperm volatiles and verbenone were tested for their ability to disrupt the response of western pine beetle, Dendroctonus brevicomis LeConte (Coleoptera: Scolytidae), to attractant-baited multiple funnel traps. Verbenone significantly reduced attraction; however, no difference was observed between 4 and
Paroma Mitra et al.
Pest management science, 77(1), 285-299 (2020-07-23)
The viviparous aphid Aphis craccivora Koch (Hemiptera: Aphididae) is a serious threat to the crop yield of Lathyrus sativus L. (Fabaceae), commonly known as grass pea. The synthetic insecticides applied to control this insect pest are not safe for the
Kye Chung Park et al.
Journal of insect physiology, 78, 33-46 (2015-05-06)
Phytophagous insects detect volatile compounds produced by host and non-host plants, using species-specific sets of olfactory receptor neurons (ORNs). To investigate the relationship between the range of host plants and the profile of ORNs, single sensillum recordings were carried out

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service