Skip to Content
MilliporeSigma
All Photos(1)

Documents

W375705

Sigma-Aldrich

Ethyl methyl p-tolylglycidate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O3
CAS Number:
Molecular Weight:
220.26
FEMA Number:
3757
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

application(s)

flavors and fragrances

InChI

1S/C13H16O3/c1-4-15-12(14)13(3)11(16-13)10-7-5-9(2)6-8-10/h5-8,11H,4H2,1-3H3

InChI key

MHZDYSAIPCCUNT-UHFFFAOYSA-N

Application

Cherry almond, vanilla, and tropical fruit nuances

Features and Benefits

Odor: Floral, green, fruity, sweet, berry and cherry-like with powdery nuances.
Taste: @ 25 PPM. Sweet, floral, fruity and cherry with almond and vanilla nuances.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 21, 52-52 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service