Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W365106

Sigma-Aldrich

6-Acetoxydihydrotheaspirane

≥90%, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O3
CAS Number:
Molecular Weight:
254.37
FEMA Number:
3651
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.087
Pricing and availability is not currently available.

assay

≥90%

mp

50-53 °C (lit.)

SMILES string

CC1CCC2(O1)C(C)(C)CCCC2(C)OC(C)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service