Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W342505

Sigma-Aldrich

Cycloheptadec-9-en-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30O
CAS Number:
Molecular Weight:
250.42
FEMA Number:
3425
EC Number:
Council of Europe no.:
11744
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.11

biological source

synthetic

Quality Level

application(s)

flavors and fragrances

SMILES string

O=C1CCCCCCCC=CCCCCCCC1

InChI

1S/C17H30O/c18-17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-17/h1-2H,3-16H2/b2-1-

InChI key

ZKVZSBSZTMPBQR-UPHRSURJSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of civetone with rat liver microsomal cytochrome P-450 and steroidogenic enzymes.
M G Rowlands et al.
Biochemical pharmacology, 37(11), 2291-2293 (1988-06-01)
Hisahiro Hagiwara et al.
Natural product communications, 7(7), 913-915 (2012-08-23)
The synthesis of (Z)-civetone (1) is described starting from oleic acid (5) via a series of reactions, intermolecular olefin self-metathesis, bromination/dehydrobromination into acetylene, semi-hydrogenation and intramolecular Dieckmann macrocyclization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service