About This Item
Recommended Products
biological source
synthetic
assay
≥99%
refractive index
n20/D 1.388 (lit.)
bp
121 °C (lit.)
density
0.869 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
SMILES string
CC(C)OC(=O)C(C)C
InChI
1S/C7H14O2/c1-5(2)7(8)9-6(3)4/h5-6H,1-4H3
InChI key
WVRPFQGZHKZCEB-UHFFFAOYSA-N
Features and Benefits
signalword
Danger
hcodes
Hazard Classifications
Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
62.6 °F - closed cup
flash_point_c
17 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service