Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W281719

Sigma-Aldrich

Onion oil, Dutch

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2817
UNSPSC Code:
12164502
NACRES:
NA.21
organoleptic:
onion
grade:
Kosher
biological source:
Allium cepa L.
food allergen:
no known allergens

biological source

Allium cepa L.

Quality Level

grade

Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 182.20

origin

The Netherlands origin

refractive index

n20/D 1.5534

bp

115-238 °C

density

1.068 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

onion

InChI

1S/C7H15NO2.ClH/c1-4-5(2)6(8)7(9)10-3;/h5-6H,4,8H2,1-3H3;1H

InChI key

GGTBEWGOPAFTTH-UHFFFAOYSA-N

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

105.0 °F - closed cup

flash_point_c

40.56 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service