Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W258709

Sigma-Aldrich

5-Hydroxy-4-octanone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)COCH2CH2CH3
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2587
EC Number:
Council of Europe no.:
2045
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.065
Pricing and availability is not currently available.

grade

Halal

assay

95%

refractive index

n20/D 1.4315 (lit.)

bp

80-82 °C/10 mmHg (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

CCCC(O)C(=O)CCC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service