Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W221800

Butyl isovalerate

natural, 98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COO(CH2)3CH3
CAS Number:
Molecular Weight:
158.24
Beilstein/REAXYS Number:
1752803
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Pricing and availability is not currently available.

grade

FG
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

assay

98%

refractive index

n20/D 1.409 (lit.)
n20/D 1.409

bp

175 °C (lit.)

density

0.857 g/mL at 25 °C
0.858 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)CC(C)C

InChI

1S/C9H18O2/c1-4-5-6-11-9(10)7-8(2)3/h8H,4-7H2,1-3H3

InChI key

AYWJSCLAAPJZEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service