About This Item
Recommended Products
biological source
synthetic
grade
Kosher
reg. compliance
FDA 21 CFR 172.515
assay
≥99%
refractive index
n20/D 1.41 (lit.)
bp
123-125 °C (lit.)
density
0.913 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
SMILES string
CCC(=O)OCC=C
InChI
1S/C6H10O2/c1-3-5-8-6(7)4-2/h3H,1,4-5H2,2H3
InChI key
XRFWKHVQMACVTA-UHFFFAOYSA-N
Other Notes
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
77.0 °F - closed cup
flash_point_c
25 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service