Skip to Content
MilliporeSigma
All Photos(1)

Documents

SPC00025

Sigma-Aldrich

tert-Butyl 3-hydroxy-3-methylazetidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

OC1(C)CN(S(C2=CC=C(C)C=C2)(=O)=O)C1

InChI

1S/C11H15NO3S/c1-9-3-5-10(6-4-9)16(14,15)12-7-11(2,13)8-12/h3-6,13H,7-8H2,1-2H3

InChI key

IDDANVBGBWXLHL-UHFFFAOYSA-N

Other Notes

Product of SpiroChem

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service