Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

R1609

Sigma-Aldrich

Sodium rhodizonate dibasic

97%

Synonym(s):

3,4,5,6-Tetraoxocyclohexene-1,2-diol disodium salt, Rhodizonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Na2O6
CAS Number:
Molecular Weight:
214.04
Beilstein/REAXYS Number:
5181379
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Product R1609 is not currently sold in your country. Contact Technical Service

Quality Level

assay

97%

technique(s)

titration: suitable

mp

>300 °C (lit.)

SMILES string

[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O

InChI

1S/C6H2O6.2Na/c7-1-2(8)4(10)6(12)5(11)3(1)9;;/h7-8H;;/q;2*+1/p-2

Inchi Key

SXWPCWBFJXDMAI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium rhodizonate dibasic can be employed as a reactant in the synthesis of:
  • Dihydroxyquinone carbonate by cyclocondensation reaction with phosgene.[1]
  • Zwitterionic systems by condensation of heteroaryl-boronic acids.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Guendalina Gentile et al.
Forensic science international, 316, 110474-110474 (2020-09-04)
In this study we have evaluated the detectability of lead deriving from Gunshot Residues (GSR) on human bone using colorimetric techniques as well as the possible interferences of decalcification on the stainability of these particles. 22 well preserved cadavers were
Preparation and Diels-Alder Cycloadditions of Substituted Vinylene Carbonates (1, 3-Dioxol-2-ones) and Related Compounds.
Jung ME, et al.
Heterocycles, 28(1), 93-97 (1989)
Zwitterionic Systems Obtained by Condensation of Heteroaryl-Boronic Acids and Rhodizonic Acid
Fornasari L, et al.
European Journal of Organic Chemistry, 2019(7), 1574-1582 (2019)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service