Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD11087

Sigma-Aldrich

m-dPEG ®13-TFP ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$285.00
1000 MG
$979.00

$285.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$285.00
1000 MG
$979.00

About This Item

Empirical Formula (Hill Notation):
C34H56F4O15
Molecular Weight:
780.79
UNSPSC Code:
51171641
NACRES:
NA.22

$285.00


Check Cart for Availability

Request a Bulk Order

assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

m-dPEG®₁₃-TFP ester is an amine reactive hydrophilic dPEG modifier.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service