N3804
1-Naphthaleneacetic acid
95%
Synonym(s):
α-Naphthaleneacetic acid, 1-Naphthylacetic acid, NAA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H10O2
CAS Number:
Molecular Weight:
186.21
Beilstein/REAXYS Number:
1308415
EC Number:
MDL number:
UNSPSC Code:
12352100
Recommended Products
assay
95%
SMILES string
OC(=O)Cc1cccc2ccccc12
InChI
1S/C12H10O2/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H,13,14)
InChI key
PRPINYUDVPFIRX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service