Skip to Content
MilliporeSigma
All Photos(1)

Documents

N25389

Supelco

1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt hydrate

Synonym(s):

3-Hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, Nitroso-R salt, Nitroso-R-salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ONC10H4(OH)(SO3Na)2 · xH2O
CAS Number:
Molecular Weight:
377.26 (anhydrous basis)
Beilstein/REAXYS Number:
3643930
EC Number:
MDL number:
UNSPSC Code:
12000000

mp

>300 °C (lit.)

λmax

372 nm

SMILES string

O.[Na+].[Na+].Oc1c(cc2cc(ccc2c1N=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C10H7NO8S2.2Na.H2O/c12-10-8(21(17,18)19)4-5-3-6(20(14,15)16)1-2-7(5)9(10)11-13;;;/h1-4,12H,(H,14,15,16)(H,17,18,19);;;1H2/q;2*+1;/p-2

InChI key

RDFCJJHALFWNIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service