Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M50109

Sigma-Aldrich

Methyl 4-hydroxybenzoate

99%

Synonym(s):

p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
509801
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

99%

mp

125-128 °C (lit.)

SMILES string

COC(=O)c1ccc(O)cc1

InChI

1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3

InChI key

LXCFILQKKLGQFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

334.4 °F

flash_point_c

168 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

San-Yong Wang et al.
Bioorganic & medicinal chemistry letters, 17(10), 2785-2788 (2007-03-28)
Twelve allenic aromatic ethers, some of them are natural products isolated from the mangrove fungus Xylaria sp. 2508 in the South China Sea, were synthesized. Their antitumor activities against KB and KBv200 cells were determined. All these compounds demonstrated cytotoxic
Peng-Cheng Lv et al.
European journal of medicinal chemistry, 44(4), 1779-1787 (2008-06-10)
Twenty-six depsides were synthesized to screen for their antibacterial activity. All of them were reported for the first time. Their chemical structures were clearly determined by (1)H NMR, (13)C NMR, ESI mass spectra and elemental analyses, coupled with one selected
Fabiana Manservisi et al.
Reproductive toxicology (Elmsford, N.Y.), 54, 110-119 (2015-01-03)
The aim of the study is to determine whether low doses of "endocrine disrupting chemicals" (EDCs) affect the development and proliferative activity of the mammary glands (MGs). Adult parous/nulliparous female Sprague-Dawley (SD) rats were treated from post-natal day (PND) 1
Peter J McNamara et al.
Antimicrobial agents and chemotherapy, 53(5), 1898-1906 (2009-02-19)
Menstrual toxic shock syndrome is a rare but potentially life-threatening illness manifest through the actions of Staphylococcus aureus toxic shock syndrome toxin 1 (TSST-1). Previous studies have shown that tampon additives can influence staphylococcal TSST-1 production. We report here on
Markus A Lill et al.
Journal of medicinal chemistry, 48(18), 5666-5674 (2005-09-02)
We investigated the influence of induced fit of the androgen receptor binding pocket on free energies of ligand binding. On the basis of a novel alignment procedure using flexible docking, molecular dynamics simulations, and linear-interaction energy analysis, we simulated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service