Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J100036

Niobium(V) ethoxide

99.999%, electronic grade

Synonym(s):

NbOEt, Niobium ethanolate, Niobium ethylate, Niobium pentaethoxide, Pentaethoxyniobium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nb(OCH2CH3)5
CAS Number:
Molecular Weight:
318.21
Beilstein/REAXYS Number:
3678883
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

electronic grade

assay

99.999%

form

liquid

refractive index

n20/D 1.516 (lit.)

bp

142 °C/0.1 mmHg (lit.)

mp

5-6 °C (lit.)

density

1.268 g/mL at 25 °C (lit.)

SMILES string

CCO[Nb](OCC)(OCC)(OCC)OCC

InChI

1S/5C2H5O.Nb/c5*1-2-3;/h5*2H2,1H3;/q5*-1;+5

InChI key

ZTILUDNICMILKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Please inquire for quantity, pricing, and packaging options.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanchiappan Aravindan et al.
ChemSusChem, 7(7), 1858-1863 (2014-06-26)
This is the first report of the utilization of TiNb2 O7 as an insertion-type anode in a lithium-ion hybrid electrochemical capacitor (Li-HEC) along with an activated carbon (AC) counter electrode derived from a coconut shell. A simple and scalable electrospinning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service