Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7756

Sigma-Aldrich

5-Iodo-2′-deoxyuridine

98%

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)-5-iodouracil, 2′-Deoxy-5-iodouridine, 5-IUdR, IDU, IdUrd, Idoxuridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11IN2O5
CAS Number:
Molecular Weight:
354.10
Beilstein/REAXYS Number:
30397
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

optical activity

[α]21/D +29.9°, c = 1 in 1 M NaOH

mp

194 °C (lit.)

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(I)C(=O)NC2=O

InChI

1S/C9H11IN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

XQFRJNBWHJMXHO-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service