Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I27902

Sigma-Aldrich

Isoprenaline hydrochloride

98%

Synonym(s):

(±)-Isoproterenol hydrochloride, 1-(3′,4′-Dihydroxyphenyl)-2-isopropylaminoethanol hydrochloride, N-Isopropyl-DL-noradrenaline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO3 · HCl
CAS Number:
Molecular Weight:
247.72
Beilstein/REAXYS Number:
3917363
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

98%

mp

165-175 °C (dec.) (lit.)

SMILES string

Cl.CC(C)NCC(O)c1ccc(O)c(O)c1

InChI

1S/C11H17NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,11-15H,6H2,1-2H3;1H

InChI key

IROWCYIEJAOFOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

β-adrenoceptor agonist; increases cytosolic cAMP.

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron M Ring et al.
Nature, 502(7472), 575-579 (2013-09-24)
G-protein-coupled receptors (GPCRs) are integral membrane proteins that have an essential role in human physiology, yet the molecular processes through which they bind to their endogenous agonists and activate effector proteins remain poorly understood. So far, it has not been
M Morgan-Bathke et al.
Journal of dental research, 92(10), 911-917 (2013-07-26)
Autophagy is a catabolic pathway utilized to maintain a balance among the synthesis, degradation, and recycling of cellular components, thereby playing a role in cell growth, development, and homeostasis. Previous studies revealed that a conditional knockout of essential member(s) of
Michael Schwarzl et al.
Critical care medicine, 41(12), e401-e410 (2013-08-22)
We tested whether mild hypothermia impacts on circulatory and respiratory dysfunction during experimental endotoxemia. Randomized controlled prospective experimental study. Large animal facility, Medical University of Graz, Austria. Thirteen anesthetized and mechanically ventilated pigs. Lipopolysaccharide was administered for 4 hours. With
Sachin Nayyar et al.
American journal of physiology. Heart and circulatory physiology, 305(8), H1181-H1188 (2013-08-13)
QT variability (QTV) signifies repolarization lability, and increased QTV is a risk predictor for sudden cardiac death. The aim of the present study was to investigate the role of autonomic nervous system activity on QTV. This study was performed in
Roshanak Irannejad et al.
Nature, 495(7442), 534-538 (2013-03-22)
A long-held tenet of molecular pharmacology is that canonical signal transduction mediated by G-protein-coupled receptor (GPCR) coupling to heterotrimeric G proteins is confined to the plasma membrane. Evidence supporting this traditional view is based on analytical methods that provide limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service