Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I11801

Sigma-Aldrich

3-Iodo-L-tyrosine

97%

Synonym(s):

3-Monoiodo-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3-4-(OH)CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
307.09
Beilstein/REAXYS Number:
2941266
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

optical activity

[α]20/D −4°, c = 5 in 1 M HCl

mp

210 °C (dec.) (lit.)

SMILES string

N[C@@H](Cc1ccc(O)c(I)c1)C(O)=O

InChI

1S/C9H10INO3/c10-6-3-5(1-2-8(6)12)4-7(11)9(13)14/h1-3,7,12H,4,11H2,(H,13,14)/t7-/m0/s1

InChI key

UQTZMGFTRHFAAM-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Tyrosine hydroxylase inhibitor.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew G Romei et al.
Science (New York, N.Y.), 367(6473), 76-79 (2020-01-04)
Rotation around a specific bond after photoexcitation is central to vision and emerging opportunities in optogenetics, super-resolution microscopy, and photoactive molecular devices. Competing roles for steric and electrostatic effects that govern bond-specific photoisomerization have been widely discussed, the latter originating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service